Assay Details
Gene Symbol: DIAPH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083789.2 | NP_001077258.1 | 1 - 2 | 331 | 91 | |
GenBank mRNA | BC134706.1 | - | 1 - 2 | 333 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
