Assay Details
| Gene Symbol: FAM122B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083794.1 | NP_001077263.1 | 7 - 8 | 569 | 61 | |
| GenBank mRNA | BC133383.1 | - | 7 - 8 | 569 | 61 | |
More Information
Set Membership: |
|

FAM122B
Genomic Map