Assay Details
| Gene Symbol: MGC157368 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083801.2 | NP_001077270.1 | 1 - 2 | 175 | 110 | |
| GenBank mRNA | BC134733.1 | - | 1 - 2 | 176 | 110 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC134733.1, NM_001083801.2 transcripts.
Set Membership: |
|

MGC157368
Genomic Map