Assay Details
| Gene Symbol: MGC157368 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083801.2 | NP_001077270.1 | 2 - 3 | 256 | 95 | |
| GenBank mRNA | BC134733.1 | - | 2 - 3 | 257 | 95 | |
More Information
Set Membership: |
|

MGC157368
Genomic Map