Assay Details
| Gene Symbol: MGC157368 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083801.2 | NP_001077270.1 | 6 - 7 | 638 | 62 | |
| GenBank mRNA | AY427788.1 | - | 3 - 4 | 234 | 62 | |
| BC134733.1 | - | 6 - 7 | 639 | 62 | ||
| Gene Symbol: AST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038119.1 | 3 - 4 | 234 | 62 | ||
More Information
Set Membership: |
|

MGC157368
Genomic Map