Assay Details
| Gene Symbol: GARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097566.1 | NP_001091035.1 | 16 - 17 | 2109 | 66 | |
| GenBank mRNA | AY568575.1 | - | 12 - 13 | 1485 | 66 | |
| BC140482.1 | - | 16 - 17 | 2109 | 66 | ||
More Information
Set Membership: |
|

GARS1
Genomic Map