Assay Details
Gene Symbol: ST3GAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097983.1 | NP_001091452.1 | 4 - 5 | 735 | 76 | |
GenBank mRNA | AJ305086.1 | - | 5 - 6 | 679 | 76 | |
BT030479.1 | - | 4 - 5 | 735 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
