Assay Details
| Gene Symbol: S100A11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098856.1 | NP_001092326.1 | 2 - 3 | 263 | 63 | |
| GenBank mRNA | AB099012.1 | - | 2 - 3 | 215 | 63 | |
| BC142378.1 | - | 2 - 3 | 263 | 63 | ||
More Information
Set Membership: |
|

S100A11
Genomic Map