Assay Details
| Gene Symbol: FASLG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098859.2 | NP_001092329.1 | 2 - 3 | 574 | 94 | |
| GenBank mRNA | AB035802.1 | - | 1 - 2 | 55 | 94 | |
| BC146056.1 | - | 2 - 3 | 576 | 94 | ||
| JN380921.1 | - | 2 - 3 | 385 | 94 | ||
More Information
Set Membership: |
|

FASLG
Genomic Map