Assay Details
| Gene Symbol: LZTS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098862.2 | NP_001092332.1 | 3 - 4 | 1195 | 104 | |
| GenBank mRNA | BC142533.1 | - | 3 - 4 | 1196 | 104 | |
More Information
Set Membership: |
|

LZTS2
Genomic Map