Assay Details
| Gene Symbol: WNT2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099363.2 | NP_001092833.1 | 1 - 2 | 459 | 55 | |
| GenBank mRNA | BC146036.1 | - | 1 - 2 | 460 | 55 | |
More Information
Set Membership: |
|

WNT2B
Genomic Map