Assay Details
| Gene Symbol: ACOT13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101845.2 | NP_001095315.1 | 1 - 2 | 193 | 100 | |
| GenBank mRNA | BC149695.1 | - | 1 - 2 | 194 | 100 | |
More Information
Set Membership: |
|

ACOT13
Genomic Map