Assay Details
Gene Symbol: ACOT13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101845.2 | NP_001095315.1 | 2 - 3 | 379 | 94 | |
GenBank mRNA | BC149695.1 | - | 2 - 3 | 380 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
