Assay Details
Gene Symbol: MSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113240.1 | NP_001106711.1 | 3 - 4 | 258 | 68 | |
NM_174113.2 | NP_776538.1 | 3 - 4 | 258 | 68 | ||
GenBank mRNA | BC142250.1 | - | 3 - 4 | 330 | 68 | |
X51689.1 | - | 2 | 233 | 68 | ||
X54183.1 | - | 247 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
