Assay Details
| Gene Symbol: PRSS42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114626.2 | NP_001108098.2 | 4 - 5 | 1017 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRSS42| Gene Symbol: PRSS42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114626.2 | NP_001108098.2 | 4 - 5 | 1017 | 93 | |
Set Membership: |
|