Assay Details
Gene Symbol: CXCL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174300.2 | NP_776725.1 | 2 - 3 | 287 | 60 | |
GenBank mRNA | AF149249.1 | - | 2 - 3 | 287 | 60 | |
BC142108.1 | - | 2 - 3 | 311 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
