Assay Details
| Gene Symbol: PDIA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174333.3 | NP_776758.2 | 11 - 12 | 1431 | 64 | |
| GenBank mRNA | BC140529.1 | - | 11 - 12 | 1431 | 64 | |
| D16235.1 | - | 11 - 12 | 1426 | 64 | ||
More Information
Set Membership: |
|

PDIA3
Genomic Map