Assay Details
| Gene Symbol: ATP5F1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174684.2 | NP_777109.1 | 1 - 2 | 82 | 110 | |
| GenBank mRNA | BC116059.1 | - | 1 - 2 | 96 | 110 | |
| M22465.1 | - | 1 - 2 | 82 | 110 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC116059.1, NM_174684.2 transcripts.
Set Membership: |
|

ATP5F1A
Genomic Map