Assay Details
| Gene Symbol: CD38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175798.3 | NP_786992.1 | 2 - 3 | 473 | 62 | |
| GenBank mRNA | AF101437.1 | - | 2 - 3 | 422 | 62 | |
| BC126708.1 | - | 2 - 3 | 473 | 62 | ||
More Information
Set Membership: |
|

CD38
Genomic Map