Assay Details
| Gene Symbol: ENDOG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175823.3 | NP_787017.1 | 2 - 3 | 832 | 66 | |
| GenBank mRNA | BC134634.1 | - | 2 - 3 | 832 | 66 | |
| X72802.1 | - | 634 | 66 | |||
More Information
Set Membership: |
|

ENDOG
Genomic Map