Assay Details
| Gene Symbol: ACVR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176663.3 | NP_788836.2 | 5 - 6 | 820 | 61 | |
| GenBank mRNA | BC133311.1 | - | 5 - 6 | 820 | 61 | |
| U58095.1 | - | 5 - 6 | 775 | 61 | ||
More Information
Set Membership: |
|

ACVR1
Genomic Map