Assay Details
| Gene Symbol: FNTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177498.3 | NP_803464.2 | 1 - 2 | 205 | 80 | |
| GenBank mRNA | AV602029.1 | - | 1 - 2 | 96 | 80 | |
| BC112662.1 | - | 1 - 2 | 199 | 80 | ||
| CF767073.1 | - | 1 - 2 | 205 | 80 | ||
| M74083.1 | - | 1 - 2 | 89 | 80 | ||
More Information
Set Membership: |
|

FNTA
Genomic Map