Assay Details
Gene Symbol: SLC3A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024488.2 | NP_001019659.2 | 2 - 3 | 733 | 73 | |
GenBank mRNA | BC102420.1 | - | 1 - 2 | 564 | 73 | |
BC123690.1 | - | 2 - 3 | 604 | 73 | ||
BT020715.1 | - | 2 - 3 | 681 | 73 | ||
BT021541.1 | - | 2 - 3 | 682 | 73 | ||
DV919154.1 | - | 2 - 3 | 803 | 73 | ||
EE364739.1 | - | 1 - 2 | 504 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
