Assay Details
| Gene Symbol: SLA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024533.2 | NP_001019704.2 | 5 - 6 | 526 | 69 | |
| GenBank mRNA | BC119980.1 | - | 5 - 6 | 526 | 69 | |
| BT021847.1 | - | 4 - 5 | 431 | 69 | ||
More Information
Set Membership: |
|

SLA
Genomic Map