Assay Details
| Gene Symbol: HEPACAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031759.2 | NP_001026929.2 | 5 - 6 | 911 | 137 | |
| GenBank mRNA | BC123427.1 | - | 5 - 6 | 911 | 137 | |
More Information
Set Membership: |
|

HEPACAM
Genomic Map