Assay Details
Gene Symbol: HEPACAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031759.2 | NP_001026929.2 | 6 - 7 | 980 | 158 | |
GenBank mRNA | BC123427.1 | - | 6 - 7 | 980 | 158 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
