Assay Details
Gene Symbol: KLF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035271.4 | NP_001030348.2 | 2 - 3 | 891 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
