Assay Details
Gene Symbol: CCDC82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046094.2 | NP_001039559.2 | 8 - 9 | 1675 | 82 | |
GenBank mRNA | AM006561.1 | - | 5 - 6 | 804 | 82 | |
CK974991.1 | - | 2 - 3 | 449 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
