Assay Details
Gene Symbol: LTC4S | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046098.2 | NP_001039563.2 | 4 - 5 | 383 | 71 | |
GenBank mRNA | BC111676.1 | - | 4 - 5 | 384 | 71 | |
DV905069.1 | - | 4 - 5 | 326 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
