Assay Details
| Gene Symbol: GSTO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075214.2 | NP_001068682.2 | 5 - 6 | 647 | 78 | |
| GenBank mRNA | BC120211.1 | - | 5 - 6 | 633 | 78 | |
| DV852688.1 | - | 3 - 4 | 382 | 78 | ||
More Information
Set Membership: |
|

GSTO1
Genomic Map