Assay Details
| Gene Symbol: ZNF180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075644.2 | NP_001069112.2 | 4 - 5 | 269 | 101 | |
| GenBank mRNA | BC149088.1 | - | 4 - 5 | 269 | 101 | |
| BT026551.1 | - | 3 - 4 | 382 | 101 | ||
More Information
Set Membership: |
|

ZNF180
Genomic Map