Assay Details
Gene Symbol: PREB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076875.2 | NP_001070343.2 | 1 - 2 | 361 | 67 | |
GenBank mRNA | BC123551.1 | - | 1 - 2 | 362 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
