Assay Details
| Gene Symbol: ATP6V0E2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097574.2 | NP_001091043.1 | 3 - 4 | 340 | 86 | |
| GenBank mRNA | BC112699.1 | - | 3 - 4 | 341 | 86 | |
More Information
Set Membership: |
|

ATP6V0E2
Genomic Map