Assay Details
| Gene Symbol: PITX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097991.1 | NP_001091460.1 | 2 - 3 | 634 | 65 | |
| GenBank mRNA | BC140579.1 | - | 2 - 3 | 634 | 65 | |
| KT336363.1 | - | 2 - 3 | 409 | 65 | ||
More Information
Set Membership: |
|

PITX2
Genomic Map