Assay Details
| Gene Symbol: NRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098002.2 | NP_001091471.1 | 9 - 10 | 1238 | 56 | |
| GenBank mRNA | BC140662.1 | - | 9 - 10 | 1239 | 56 | |
More Information
Set Membership: |
|

NRF1
Genomic Map