Assay Details
| Gene Symbol: COX10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098013.1 | NP_001091482.1 | 1 - 2 | 120 | 81 | |
| GenBank mRNA | BC140517.1 | - | 1 - 2 | 120 | 81 | |
More Information
Set Membership: |
|

COX10
Genomic Map