Assay Details
Gene Symbol: PUM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098030.1 | NP_001091499.1 | 12 - 13 | 1239 | 94 | |
GenBank mRNA | BC133332.1 | - | 12 - 13 | 1239 | 94 | |
BT030563.1 | - | 12 - 13 | 1227 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
