Assay Details
Gene Symbol: PUM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098030.1 | NP_001091499.1 | 15 - 16 | 1682 | 72 | |
GenBank mRNA | BC133332.1 | - | 15 - 16 | 1682 | 72 | |
BT030563.1 | - | 15 - 16 | 1670 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
