Assay Details
| Gene Symbol: CLMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098043.1 | NP_001091512.1 | 2 - 3 | 486 | 61 | |
| GenBank mRNA | BC140573.1 | - | 2 - 3 | 486 | 61 | |
More Information
Set Membership: |
|

CLMP
Genomic Map