Assay Details
| Gene Symbol: COPRS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098894.2 | NP_001092364.1 | 3 - 4 | 428 | 76 | |
| GenBank mRNA | BC142058.1 | - | 3 - 4 | 429 | 76 | |
More Information
Set Membership: |
|

COPRS
Genomic Map