Assay Details
| Gene Symbol: CAPSL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098899.2 | NP_001092369.1 | 2 - 3 | 175 | 112 | |
| GenBank mRNA | BC146156.1 | - | 2 - 3 | 176 | 112 | |
More Information
Set Membership: |
|

CAPSL
Genomic Map