Assay Details
| Gene Symbol: FRS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098918.2 | NP_001092388.1 | 5 - 6 | 623 | 74 | |
| GenBank mRNA | BC146158.1 | - | 5 - 6 | 624 | 74 | |
More Information
Set Membership: |
|

FRS3
Genomic Map