Assay Details
| Gene Symbol: TRAPPC2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098920.1 | NP_001092390.1 | 4 - 5 | 379 | 151 | |
| GenBank mRNA | BC146228.1 | - | 4 - 5 | 379 | 151 | |
More Information
Set Membership: |
|

TRAPPC2L
Genomic Map