Assay Details
Gene Symbol: NUP35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098940.1 | NP_001092410.1 | 4 - 5 | 447 | 75 | |
GenBank mRNA | BC142122.1 | - | 4 - 5 | 447 | 75 | |
BC149564.1 | - | 4 - 5 | 462 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
