Assay Details
| Gene Symbol: MLYCD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098946.1 | NP_001092416.1 | 4 - 5 | 983 | 100 | |
| GenBank mRNA | BC142049.1 | - | 4 - 5 | 983 | 100 | |
More Information
Set Membership: |
|

MLYCD
Genomic Map