Assay Details
| Gene Symbol: MAPK12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098953.1 | NP_001092423.1 | 3 - 4 | 430 | 62 | |
| GenBank mRNA | BC142157.1 | - | 3 - 4 | 430 | 62 | |
More Information
Set Membership: |
|

MAPK12
Genomic Map