Assay Details
| Gene Symbol: MLEC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098983.1 | NP_001092453.1 | 4 - 5 | 769 | 65 | |
| GenBank mRNA | BC110237.1 | - | 4 - 5 | 764 | 65 | |
| BC146164.1 | - | 4 - 5 | 769 | 65 | ||
More Information
Set Membership: |
|

MLEC
Genomic Map