Assay Details
| Gene Symbol: SLC6A13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099027.1 | NP_001092497.1 | 5 - 6 | 618 | 59 | |
| GenBank mRNA | BC142276.1 | - | 5 - 6 | 618 | 59 | |
More Information
Set Membership: |
|

SLC6A13
Genomic Map