Assay Details
| Gene Symbol: TMEM189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100315.1 | NP_001093785.1 | 2 - 3 | 266 | 56 | |
| GenBank mRNA | BC148012.1 | - | 2 - 3 | 266 | 56 | |
| BC149405.1 | - | 2 - 3 | 305 | 56 | ||
More Information
Set Membership: |
|

TMEM189
Genomic Map