Assay Details
| Gene Symbol: FKBP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100330.2 | NP_001093800.1 | 1 - 2 | 295 | 80 | |
| GenBank mRNA | BC149352.1 | - | 1 - 2 | 297 | 80 | |
More Information
Set Membership: |
|

FKBP7
Genomic Map